ACES 99+% Biochemistry
Dodavatel: Thermo Fisher Scientific
Synonyms:
ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
327671000
327670010
327672500
ACRO327671000EA
5120
CZK
ACRO327671000
ACRO327670010
ACRO327672500
ACES 99+% Biochemistry
ACES
Vzorec:
H₂NCOCH₂NHCH₂CH₂SO₃H MW: 182.2 Bod tání: 270…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008030 CAS číslo: 7365-82-4 EINECS: 230-908-4 Merck Index: 13,00037 |
|
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
Heavy metals | ≤5 ppm |
UV | at 280 nm A: ≤0.02 |
pH | 3.6 to 4.4 (1 % at 25°C) |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...