Močovina, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
MQ200
15604-1KG
15604-2.5KG
15604-250G
15604-25KG
15604-20KG
SIAL15604-1KGEA
1130
CZK
SIAL15604-1KG
SIAL15604-2.5KG
SIAL15604-250G
SIAL15604-25KG
SIAL15604-20KG
Močovina, Sigma-Aldrich®
Močovina
Vzorec:
NH₂CONH₂ MW: 60.06 Bod varu: 197 Bod tání: 132.7 Hustota: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS číslo: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 13,09935 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...