ADA, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
M-Clarity™ quality level = MQ400
A9883-100G
A9883-1KG
A9883-250G
A9883-25G
SIALA9883-100GEA
9920
CZK
SIALA9883-100G
SIALA9883-1KG
SIALA9883-250G
SIALA9883-25G
ADA, Sigma-Aldrich®
ADA
Vzorec:
H₂NC(O)CH₂N(CH₂COOH)₂ MW: 190,16 g/mol Bod tání: 217…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008031 CAS číslo: 26239-55-4 EINECS: 247-530-0 Merck Index: 13,00148 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...