ADA 99% for biochemistry
Dodavatel: Thermo Scientific
Synonyms:
2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
228010250
228011000
ACRO228010250EA
923
CZK
ACRO228010250
ACRO228011000
ADA 99% for biochemistry
ADA
Vzorec:
H₂NC(O)CH₂N(CH₂COOH)₂ MW: 190,16 g/mol Bod tání: 219 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00008031 CAS číslo: 26239-55-4 EINECS: 247-530-0 Merck Index: 13,00148 |
Výsledky specifikačního testu
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (110°C, 5 hrs) |
Sulfated ash | ≤0.1 % |
UV | at 300 nm A: ≤0.08 |
Appearance of solution | (9% soln. 1 M NaOH) clear, colorless |
Titration with NaOH | ≥98.5 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...