Formamid 99+% for spectrophotometry
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Carbamaldehyde
44066.AE
44066.AP
44066.AY
44066.AEEA
1740
CZK
44066.AE
44066.AP
44066.AY
Formamid 99+% for spectrophotometry
Formamid
Purity: 99+%
CAS No.: 75-12-7
Molecular Formula: CH3NO
Formula Weight: 45.04
Flash Point: 154°(310°F)
Physical Form: Liquid
Density: 1.134
Refractive Index: 1.4475
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Ammonia-like
Melting Point: 2°
Boiling Point: 210°
Beilstein Reference: 505995
Merck Reference: 14,4237
MDL No.: MFCD00007941
Vzorec:
HCONH₂ MW: 45.04 Bod varu: 210 °C Bod tání: 2 Hustota: 1.134 Bod vzplanutí: 154 °C (310 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007941 CAS číslo: 75-12-7 EINECS: 200-842-0 Merck Index: 14,4237 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...