N,N-Dimethylformamid ≥99.7% for HPLC
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
22915.K2
22915.M1
22915.Y0
22915.K2EA
3120
CZK
22915.K2
22915.M1
22915.Y0
N,N-Dimethylformamid ≥99.7% for HPLC
N,N-Dimethylformamid
Purity: 99.7+%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Physical Form: Liquid
Density: 0.944
Refractive Index: 1.4310
Odour: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Vzorec:
HCON(CH₃)₂ Bod varu: 153 °C Bod tání: –61 °C Hustota: 0,944 Bod vzplanutí: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS číslo: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 14,3243 |