Tisk…

N,N-Dimethylacetamid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

M-Clarity™ quality level = MQ100

185884-500ML 185884-6X500ML
SIAL185884-500MLEA 2670 CZK
SIAL185884-500ML SIAL185884-6X500ML
N,N-Dimethylacetamid, Sigma-Aldrich®
N,N-Dimethylacetamid
Vzorec: CH₃CON(CH₃)₂
MW: 87,12 g/mol
Bod varu: 166,1 °C (1013 hPa)
Bod tání: –20 °C
Hustota: 0,937 g/cm³ (20 °C)
Bod vzplanutí: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS číslo: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR