Tisk…

Stearoyl chloride, Sigma-Aldrich®

Dodavatel: Merck

85730-100ML 85730-500ML
SIAL85730-100MLEA 2330 CZK
SIAL85730-100ML SIAL85730-500ML
Stearoyl chloride, Sigma-Aldrich®
Stearoyl chloride
Vzorec: C₁₈H₃₅ClO
MW: 302.93
Bod varu: 348
Bod tání: 164…167 °C
Hustota: 0.9007 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000744
CAS číslo: 112-76-5
EINECS: 204-004-5
UN: 3265

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR