Stearoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
171158-25G
171158-5G
SIAL171158-25GEA
8800
CZK
SIAL171158-25G
SIAL171158-5G
Stearoyl chloride, Sigma-Aldrich®
Stearoyl chloride
Vzorec:
C₁₈H₃₅ClO MW: 302.93 Bod varu: 348 Bod tání: 164…167 °C Hustota: 0.9007 Storage Temperature: Ambient |
MDL Number:
MFCD00000744 CAS číslo: 112-76-5 EINECS: 204-004-5 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...