Oxalyl dibromide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
113034-100G
113034-25G
SIAL113034-100GEA
11300
CZK
SIAL113034-100G
SIAL113034-25G
Oxalyl dibromide, Sigma-Aldrich®
Oxalyl dibromide
Vzorec:
C₂Br₂O₂ Bod varu: 118 °C (1013 hPa) Hustota: 2,3 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000113 CAS číslo: 15219-34-8 EINECS: 239-271-7 UN: 2922 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...