Tisk…

Aluminium acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

M-Clarity™ quality level = MQ200

674753-25G 674753-5G
SIAL674753-25GEA 7750 CZK
SIAL674753-25G SIAL674753-5G
Aluminium acetylacetonate, Sigma-Aldrich®
Aluminium acetylacetonate
Vzorec: C₁₅H₂₁AlO₆
MW: 324,31 g/mol
Bod varu: 315 °C (1013 hPa)
Bod tání: 192…195 °C
Hustota: 1,204 g/cm³ (20 °C)
MDL Number: MFCD00000013
CAS číslo: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR