Aluminium acetylacetonate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone
M-Clarity™ quality level = MQ200
208248-100G
208248-500G
SIAL208248-100GEA
1310
CZK
SIAL208248-100G
SIAL208248-500G
Aluminium acetylacetonate, Sigma-Aldrich®
Aluminium acetylacetonate
Vzorec:
C₁₅H₂₁AlO₆ MW: 324,31 g/mol Bod varu: 315 °C (1013 hPa) Bod tání: 192…195 °C Hustota: 1,204 g/cm³ (20 °C) |
MDL Number:
MFCD00000013 CAS číslo: 13963-57-0 EINECS: 237-741-6 UN: 3467 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...