Tisk…

Aluminium acetylacetonate ≥99.995% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone

44538.06 44538.14 44538.22
44538.06EA 1920 CZK
44538.06 44538.14 44538.22
Aluminium acetylacetonate ≥99.995% (metals basis)
Aluminium acetylacetonate

Purity: 99.995+% CAS No.: 13963-57-0 Molecular Formula: C15H21AlO6 Formula Weight: 324.31 Physical Form: Powder Density: 1.27 Melting Point: 192-194° Boiling Point: 314-316° Beilstein Reference: 4157942 UN No.: 3467 MDL No.: MFCD00000013

Vzorec: C₁₅H₂₁AlO₆
MW: 324.31
Bod varu: 314…316 °C
Bod tání: 192…194 °C
Hustota: 1.27
Storage Temperature: Ambient
MDL Number: MFCD00000013
CAS číslo: 13963-57-0
EINECS: 237-741-6
UN: 3467
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR