Diacethyl, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,3-Butanedione
M-Clarity™ quality level = MQ100
B85307-100ML
B85307-500ML
B85307-5ML
SIALB85307-100MLEA
1400
CZK
SIALB85307-100ML
SIALB85307-500ML
SIALB85307-5ML
Diacethyl, Sigma-Aldrich®
Diacethyl
Vzorec:
C₄H₆O₂ MW: 86,09 g/mol Bod varu: 190 °C (1013 hPa) Bod tání: –2 °C Hustota: 0,9904 g/cm³ (20 °C) Bod vzplanutí: 9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008756 CAS číslo: 431-03-8 EINECS: 207-069-8 UN: 2346 ADR: 3,II Merck Index: 13,02985 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...