Tisk…

3-Mercaptobutanone, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: 2-Mercapto-3-butanone, 3-Mercapto-2-butanone

M-Clarity™ quality level = MQ300

W329800-100G-K W329800-1KG-K
SIALW329800-100G-KEA 6570 CZK
SIALW329800-100G-K SIALW329800-1KG-K
3-Mercaptobutanone, Sigma-Aldrich®
3-Mercaptobutanone
Vzorec: C₄H₈OS
MW: 104.17
MDL Number: MFCD00085206
CAS číslo: 40789-98-8
EINECS: 255-082-2
UN: 1224
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR