Diphenyl carbonate ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
A12362.0B
A12362.0I
A12362.30
A12362.0BEA
2260
CZK
A12362.0B
A12362.0I
A12362.30
Diphenyl carbonate ≥99%
Diphenyl carbonate
Purity: 99%
CAS No.: 102-09-0
Molecular Formula: C13H10O3
Formula Weight: 214.22
Flash Point: 168°(334°F)
Odour: Mild
Boiling Point: 301-302°
Beilstein Reference: 1074863
Merck Reference: 14,3602
MDL No.: MFCD00003037
Vzorec:
C₁₃H₁₀O₃ MW: 214,22 g/mol Bod varu: 301…302 °C Bod tání: 76…81 °C Hustota: 1,300 Bod vzplanutí: 168 °C (334 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003037 CAS číslo: 102-09-0 EINECS: 203-005-8 Merck Index: 14,3602 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...