Tisk…

Oxalacetic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

O4126-5G O4126-100G O4126-1G O4126-25G
SIALO4126-5GEA 4760 CZK
SIALO4126-5G SIALO4126-100G SIALO4126-1G SIALO4126-25G
Oxalacetic acid, Sigma-Aldrich®
Oxalacetic acid
Vzorec: C₄H₄O₅
Bod varu: 310 °C (1013 hPa)
Bod tání: 182…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002592
CAS číslo: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Merck Index: 12,07041

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR