Oxalacetic acid ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid
A12739.06
A12739.14
A12739.22
A12739.06EA
3010
CZK
A12739.06
A12739.14
A12739.22
Oxalacetic acid ≥97%
Oxalacetic acid
Purity: 97%
CAS No.: 328-42-7
Molecular Formula: C4H4O5
Formula Weight: 132.07
Storage Temperature: -30°C to -10°C
Flash Point: 88°(190°F)
Melting Point: 161° dec.
Beilstein Reference: 1705475
Merck Reference: 14,6909
UN No.: 3261
MDL No.: MFCD00002592
Vzorec:
C₄H₄O₅ MW: 132,07 g/mol Bod varu: 310 °C (1013 hPa) Bod tání: 161 °C (rozkl.) Bod vzplanutí: 88 °C (190 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00002592 CAS číslo: 328-42-7 EINECS: 206-329-8 UN: 3261 ADR: 8,II Merck Index: 14,6909 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...