Tisk…

Oxalacetic acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid

15789.04 15789.09
15789.04EA 1790 CZK
15789.04 15789.09
Oxalacetic acid ≥98%
Oxalacetic acid

Purity: 98+% CAS No.: 328-42-7 Molecular Formula: C4H4O5 Formula Weight: 132.07 Flash Point: 88°(190°F) Physical Form: Powder Melting Point: 161° dec. Beilstein Reference: 1705475 Merck Reference: 14,6909 UN No.: 3261 MDL No.: MFCD00002592

Vzorec: C₄H₄O₅
MW: 132,07 g/mol
Bod varu: 310 °C (1013 hPa)
Bod tání: 161 °C (rozkl.)
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002592
CAS číslo: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Merck Index: 14,6909

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR