Oxalacetic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Oxaloacetic acid, 2-Succinic acid, Oxalacetic acid
416600250
416600050
416601000
ACRO416600250EA
5920
CZK
ACRO416600250
ACRO416600050
ACRO416601000
Oxalacetic acid 98%
Oxalacetic acid
Vzorec:
C₄H₄O₅ MW: 132.07 Bod varu: 310 Bod tání: 161 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002592 CAS číslo: 328-42-7 EINECS: 206-329-8 UN: 3261 ADR: 8 II Merck Index: 12,07041 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...