Kyselina 2-oxoglutarová, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid
M-Clarity™ quality level = MQ200
K1128-100G
K1128-25G
K1128-5G
SIALK1128-100GEA
4570
CZK
SIALK1128-100G
SIALK1128-25G
SIALK1128-5G
Kyselina 2-oxoglutarová, Sigma-Aldrich®
Kyselina 2-oxoglutarová
Vzorec:
HOOCCH₂CH₂COCOOH MW: 146,1 g/mol Bod varu: 323 °C (1013 hPa) Bod tání: 112…116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004165 CAS číslo: 328-50-7 EINECS: 206-330-3 Merck Index: 13,05320 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...