Kyselina pyrohroznová ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
A13875.0E
A13875.22
A13875.36
A13875.0EEA
23890
CZK
A13875.0E
A13875.22
A13875.36
Kyselina pyrohroznová ≥98%
Kyselina pyrohroznová
Purity: 98%
CAS No.: 127-17-3
Molecular Formula: C3H4O3
Formula Weight: 88.06
Storage Temperature: 2°C to 8°C
Flash Point: 83°(181°F)
Physical Form: Liquid
Density: 1.267
Refractive Index: 1.4295
Appearance: Colourless to yellow
Odour: Pungent
pH: 1.2
Melting Point: 11-12°
Boiling Point: 164-166°
Beilstein Reference: 506211
Merck Reference: 14,8021
UN No.: 3265
MDL No.: MFCD00002585
Vzorec:
C₃H₄O₃ MW: 88,06 g/mol Bod varu: 164…166 °C Bod tání: 11…12 °C Hustota: 1,267 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002585 CAS číslo: 127-17-3 EINECS: 204-824-3 UN: 3265 ADR: 8,II Merck Index: 14,8021 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...