Tisk…

Ethyl pyruvate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester

M-Clarity™ quality level = MQ300

W245712-100G-K W245712-1KG-K W245712-5KG-K
SIALW245712-100G-KEA 0 CZK
SIALW245712-100G-K SIALW245712-1KG-K SIALW245712-5KG-K
Ethyl pyruvate, Sigma-Aldrich®
Ethyl pyruvate
Vzorec: C₅H₈O₃
Bod varu: 144…147 °C (1013 hPa)
Bod tání: –50 °C
Hustota: 1,05 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009123
CAS číslo: 617-35-6
EINECS: 210-511-2
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR