Ethyl pyruvate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A15045.18
A15045.30
A15045.36
A15045.18EA
1110
CZK
A15045.18
A15045.30
A15045.36
Ethyl pyruvate ≥98%
Ethyl pyruvate
Purity: 98%
CAS No.: 617-35-6
Molecular Formula: C5H8O3
Formula Weight: 116.12
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4050
Odour: Pleasant
Melting Point: -58°
Boiling Point: 143-145°
Beilstein Reference: 1071466
Merck Reference: 14,8021
UN No.: 3272
MDL No.: MFCD00009123
Vzorec:
C₅H₈O₃ Bod varu: 143…145 °C Bod tání: –58 °C Hustota: 1,050 Bod vzplanutí: 45 °C (113 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009123 CAS číslo: 617-35-6 EINECS: 210-511-2 UN: 3272 ADR: 3,III Merck Index: 14,8021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...