Tisk…

Acetoacetanilide ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Oxo-N-phenyl butamide, 3-Oxo-N-phenyl butanamide

A17534.0B A17534.30
A17534.0BEA 2110 CZK
A17534.0B A17534.30
Acetoacetanilide ≥98%
Acetoacetanilide

Purity: 98+% CAS No.: 102-01-2 Molecular Formula: C6H5NHCOCH2COCH3 Formula Weight: 177.20 Flash Point: 150°(302°F) Odour: Odourless Boiling Point: 129°/24mm Beilstein Reference: 473419 Merck Reference: 14,58 MDL No.: MFCD00008780

Vzorec: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Bod varu: 129 °C (24 mmHg)
Bod tání: 83…86 °C
Hustota: 1,260
Bod vzplanutí: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008780
CAS číslo: 102-01-2
EINECS: 202-996-4
Merck Index: 14,58

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR