2-Undecanone, Sigma-Aldrich®
Dodavatel: Merck
|
|
Synonyms:
Methyl nonyl ketone, Nonyl methyl ketone
M-Clarity™ quality level = MQ100
U1303-250ML
U1303-5ML
SIALU1303-250MLEA
2300
CZK
SIALU1303-250ML
SIALU1303-5ML
2-Undecanone, Sigma-Aldrich®
2-Undecanone
Vzorec:
C₁₁H₂₂O MW: 170.3 Bod varu: 231 Bod tání: ∼12 °C Hustota: 0.83 Bod vzplanutí: 96 Storage Temperature: Ambient |
MDL Number:
MFCD00009583 CAS číslo: 112-12-9 EINECS: 203-937-5 UN: 3082 ADR: 9 III Merck Index: 12,06178 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...