Tisk…

2-Undecanone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A17659.0E A17659.22 A17659.36
A17659.0EEA 12290 CZK
A17659.0E A17659.22 A17659.36
2-Undecanone ≥98%
2-Undecanone

Purity: 98% CAS No.: 112-12-9 Molecular Formula: CH3(CH2)8COCH3 Formula Weight: 170.30 Flash Point: 89°(192°F) Refractive Index: 1.4300 Melting Point: 11-13° Boiling Point: 228° Beilstein Reference: 1749573 Merck Reference: 14,6104 UN No.: 3082 MDL No.: MFCD00009583

Vzorec: C₁₁H₂₂O
Bod varu: 228 °C
Bod tání: 11…13 °C
Hustota: 0,825
Bod vzplanutí: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009583
CAS číslo: 112-12-9
EINECS: 203-937-5
UN: 3082
ADR: 9,III
Merck Index: 14,6104

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR