2-Nonanone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyl heptyl ketone, Heptyl methyl ketone
M-Clarity™ quality level = MQ300
W278505-1KG-K
W278505-250G-K
W278505-5KG-K
SIALW278505-1KG-KEA
8310
CZK
SIALW278505-1KG-K
SIALW278505-250G-K
SIALW278505-5KG-K
2-Nonanone, Sigma-Aldrich®
2-Nonanone
Vzorec:
C₉H₁₈O MW: 142,24 g/mol Bod varu: 192 °C (1013 hPa) Bod tání: –15 °C Hustota: 0,82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009553 CAS číslo: 821-55-6 EINECS: 212-480-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...