Tisk…

Succinonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L05037.22 L05037.36
L05037.22EA 1460 CZK
L05037.22 L05037.36
Succinonitrile ≥98%
Succinonitrile

Purity: 98% CAS No.: 110-61-2 Molecular Formula: C4H4N2 Formula Weight: 80.09 Flash Point: 132°(269°F) Odour: Odourless Boiling Point: 265-267° Beilstein Reference: 1098380 Merck Reference: 14,8872 UN No.: 3439 MDL No.: MFCD00001949

Vzorec: C₄H₄N₂
Bod varu: 265…267 °C
Bod tání: 52…56 °C
Hustota: 1,023 g/cm³ (20 °C)
Bod vzplanutí: 132 °C (269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001949
CAS číslo: 110-61-2
EINECS: 203-783-9
UN: 3439
ADR: 6.1,III
Merck Index: 14,8872

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR