Tisk…

Propiononitrile ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A13203.AK A13203.AU
A13203.AKEA 1260 CZK
A13203.AK A13203.AU
Propiononitrile ≥99%
Propiononitrile

Purity: 99% CAS No.: 107-12-0 Molecular Formula: CH3CH2CN Formula Weight: 55.08 Flash Point: 6°(42°F) Refractive Index: 1.3660 Odour: Pleasant Melting Point: -92° Boiling Point: 97-98° Beilstein Reference: 773680 Merck Reference: 14,7827 UN No.: 2404 MDL No.: MFCD00001948

Vzorec: C₃H₅N
Bod varu: 97…98 °C
Bod tání: –92 °C
Hustota: 0,781
Bod vzplanutí: 6 °C (42 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS číslo: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 14,7827

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR