Tisk…

Chloroacetic anhydride ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A14015.14 A14015.22 A14015.36
A14015.14EA 1650 CZK
A14015.14 A14015.22 A14015.36
Chloroacetic anhydride ≥97%
Chloroacetic anhydride

Purity: 97% CAS No.: 541-88-8 Molecular Formula: (ClCH2CO)2O Formula Weight: 170.98 Flash Point: 143°(289°F) Melting Point: 56-59° Boiling Point: 110-112°/10mm Solubility: Freely soluble in ether, chloroform. Slightly soluble in benzene. Insoluble in cold ligroin Beilstein Reference: 774533 Merck Reference: 14,2113 UN No.: 3261 MDL No.: MFCD00000929

Vzorec: C₄H₄Cl₂O₃
MW: 170,98 g/mol
Bod varu: 110…112 °C (10 mmHg)
Bod tání: 56…59 °C
Hustota: 1,5497 g/cm³ (20 °C)
Bod vzplanutí: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000929
CAS číslo: 541-88-8
EINECS: 208-794-2
UN: 3261
ADR: 8,II
Merck Index: 14,2113

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR