Tisk…

Chloroacetaldehyde dimethyl acetal, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Chloro-2,2-dimethoxyethane

M-Clarity™ quality level = MQ100

C19406-100G C19406-500G
SIALC19406-100GEA 1070 CZK
SIALC19406-100G SIALC19406-500G
Chloroacetaldehyde dimethyl acetal, Sigma-Aldrich®
Chloroacetaldehyde dimethyl acetal
Vzorec: C₄H₉ClO₂
MW: 124,57 g/mol
Bod varu: 128 °C (1013 hPa)
Bod tání: < –73 °C
Hustota: 1,098 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000948
CAS číslo: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR