Tisk…

2-Chloroethyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Isocyanic acid 2-Chloroethyl ester

M-Clarity™ quality level = MQ100

538337-25G 538337-5G
SIAL538337-25GEA 2560 CZK
SIAL538337-25G SIAL538337-5G
2-Chloroethyl isocyanate, Sigma-Aldrich®
2-Chloroethyl isocyanate
Vzorec: C₃H₄ClNO
MW: 105,52 g/mol
Bod varu: 135 °C (1013 hPa)
Hustota: 1,24 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002043
CAS číslo: 1943-83-5
EINECS: 217-734-4
UN: 3080
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR