Tisk…

Chlorodifluoroacetic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Difluorochloroacetic acid

M-Clarity™ quality level = MQ100

C34200-100G C34200-25G
SIALC34200-100GEA 0 CZK
SIALC34200-100G SIALC34200-25G
Chlorodifluoroacetic acid, Sigma-Aldrich®
Chlorodifluoroacetic acid
Vzorec: C₂HClF₂O₂
MW: 130,48 g/mol
Bod varu: 122 °C (1013 hPa)
Hustota: 2,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004176
CAS číslo: 76-04-0
EINECS: 200-928-8
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR