Methyl chlorodifluoroacetate 98%, rods
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A18623.09
A18623.30
A18623.09EA
1170
CZK
A18623.09
A18623.30
Methyl chlorodifluoroacetate 98%, rods
Methyl chlorodifluoroacetate
Purity: 98%
CAS No.: 1514-87-0
Molecular Formula: ClCF2CO2CH3
Formula Weight: 144.50
Flash Point: 19°(66°F)
Refractive Index: 1.3500
Boiling Point: 79-81°
Beilstein Reference: 1756072
UN No.: 2924
MDL No.: MFCD00000775
Vzorec:
C₃H₃ClF₂O₂ Bod varu: 79…81 °C Hustota: 1,371 Bod vzplanutí: 19 °C (66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000775 CAS číslo: 1514-87-0 EINECS: 216-154-9 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...