Tisk…

Ethyl chlorodifluoroacetate 98%, rods

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ethyl 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic ethyl ester

A14232.14 A14232.22
A14232.14EA 2150 CZK
A14232.14 A14232.22
Ethyl chlorodifluoroacetate 98%, rods
Ethyl chlorodifluoroacetate

Purity: 98% CAS No.: 383-62-0 Molecular Formula: ClF2CCO2CH2CH3 Formula Weight: 158.53 Flash Point: 18°(64°F) Refractive Index: 1.3580 Boiling Point: 96-97° Beilstein Reference: 1761413 UN No.: 2924 MDL No.: MFCD00013662

Vzorec: C₄H₅ClF₂O₂
MW: 158,53 g/mol
Bod varu: 96…97 °C
Hustota: 1,252
Bod vzplanutí: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013662
CAS číslo: 383-62-0
EINECS: 206-850-0
UN: 2924
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR