Tisk…

Perchloroethane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hexachloroethane

MQ100

185442-5G 185442-100G 185442-500G
SIAL185442-5GEA 1270 CZK
SIAL185442-5G SIAL185442-100G SIAL185442-500G
Perchloroethane, Sigma-Aldrich®
Perchloroethane
Vzorec: C₂Cl₆
MW: 236.74
Bod varu: 189
Hustota: 2.091 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000799
CAS číslo: 67-72-1
EINECS: 200-666-4
Merck Index: 13,04698

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR