Tisk…

1,3-Dichloropropane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Trimethylendichloride

M-Clarity™ quality level = MQ200

D72204-100G D72204-25G
SIALD72204-100GEA 14540 CZK
SIALD72204-100G SIALD72204-25G
1,3-Dichloropropane, Sigma-Aldrich®
1,3-Dichloropropane
Vzorec: ClH₂CCH₂CH₂Cl
MW: 112,99 g/mol
Bod varu: 120,4 °C (1013 hPa)
Bod tání: –99,5 °C
Hustota: 1,1717 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000999
CAS číslo: 142-28-9
EINECS: 205-531-3
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR