Pelargonaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Nonanal, Nonylaldehyde, Nonanal, Nonaldehyde
M-Clarity™ quality level = MQ100
N30803-25G
N30803-100G
N30803-2G
SIALN30803-25GEA
1190
CZK
SIALN30803-25G
SIALN30803-100G
SIALN30803-2G
Pelargonaldehyde, Sigma-Aldrich®
Pelargonaldehyde
Vzorec:
C₉H₁₈O Bod varu: 190 °C (1013 hPa) Bod tání: –40 °C Hustota: 0,83 g/cm³ (20 °C) Bod vzplanutí: 76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007030 CAS číslo: 124-19-6 EINECS: 204-688-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...