Tisk…

Laurinaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W261513-100G-K W261513-1KG-K
SIALW261513-100G-KEA 2890 CZK
SIALW261513-100G-K SIALW261513-1KG-K
Laurinaldehyde, Sigma-Aldrich®
Laurinaldehyde
Vzorec: C₁₂H₂₄O
Bod varu: 242 °C (1013 hPa)
Bod tání: 44 °C
Hustota: 0,83 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007017
CAS číslo: 112-54-9
EINECS: 203-983-6
UN: 3082
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR