Caprinaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Decylaldehyde, Decanal, Decyl aldehyde
M-Clarity™ quality level = MQ300
W236217-100G-K
W236217-1KG-K
W236217-25G-K
SIALW236217-100G-KEA
3640
CZK
SIALW236217-100G-K
SIALW236217-1KG-K
SIALW236217-25G-K
Caprinaldehyde, Sigma-Aldrich®
Caprinaldehyde
Vzorec:
C₁₀H₂₀O MW: 156,27 g/mol Bod varu: 207 °C (1013 hPa) Bod tání: 7 °C Hustota: 0,83 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007031 CAS číslo: 112-31-2 EINECS: 203-957-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)