(±)-1,2-Epoxypropan, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
M-Clarity™ quality level = MQ200
110205-1L
110205-2.5L
110205-500ML
SIAL110205-1LEA
2680
CZK
SIAL110205-1L
SIAL110205-2.5L
SIAL110205-500ML
(±)-1,2-Epoxypropan, Sigma-Aldrich®
(±)-1,2-Epoxypropan
Vzorec:
C₃H₆O MW: 58.08 Bod varu: 34.3 Bod tání: –112 °C Hustota: 0.83 g/cm³ (20 °C) Bod vzplanutí: –37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005126 CAS číslo: 75-56-9 EINECS: 200-879-2 UN: 1280 ADR: 3 I Merck Index: 12,08041 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...