Tisk…

4-Bromo-1-butanol cont. varying amounts of THF, tech.

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Bromo-4-hydroxybutane

16805.03 16805.06 16805.14
16805.03EA 2260 CZK
16805.03 16805.06 16805.14
4-Bromo-1-butanol cont. varying amounts of THF, tech.
4-Bromo-1-butanol

CAS No.: 33036-62-3 Molecular Formula: C4H9BrO Formula Weight: 153.02 Storage Temperature: -30°C to -10°C Flash Point: 32°(91°F) Physical Form: Liquid Sensitivity: Moisture and Light Sensitive Appearance: Yellow Boiling Point: 63-65°/5mm UN No.: 1987 MDL No.: MFCD00015387

Vzorec: BrH₂C(CH₂)₂CH₂OH
Bod varu: 63…65 °C (5 mmHg)
Bod vzplanutí: 32 °C (91 °F)
Storage Temperature: Freezer
MDL Number: MFCD00015387
CAS číslo: 33036-62-3
EINECS: 251-355-5
UN: 1987
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR