Bromoacetaldehyde diethyl acetal, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Bromo-1,1-diethoxyethane
123986-500G
123986-100G
123986-1KG
SIAL123986-500GEA
2930
CZK
SIAL123986-500G
SIAL123986-100G
SIAL123986-1KG
Bromoacetaldehyde diethyl acetal, Sigma-Aldrich®
Bromoacetaldehyde diethyl acetal
Vzorec:
BrH₂CCH(OCH₂CH₃)₂ MW: 197,07 g/mol Bod varu: 66 °C (1013 hPa) Hustota: 1,27 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000214 CAS číslo: 2032-35-1 EINECS: 217-989-1 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...