Tisk…

Ethyl bromodifluoroacetate 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ethyl 2-bromo-2,2-difluoroacetate, Bromodifluoroacetic ethyl ester

305800050 305800010 305800250
ACRO305800050EA 2250 CZK
ACRO305800050 ACRO305800010 ACRO305800250
Ethyl bromodifluoroacetate 98%
Ethyl bromodifluoroacetate
Vzorec: BrF₂CCOOCH₂CH₃
MW: 202,98 g/mol
Bod varu: 112 °C (760 mmHg)
Hustota: 1,583 g/cm³
Bod vzplanutí: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00042069
CAS číslo: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3890 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR