6-Bromocaproic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15761.14
A15761.22
A15761.36
A15761.14EA
2070
CZK
A15761.14
A15761.22
A15761.36
6-Bromocaproic acid ≥98%
6-Bromocaproic acid
Purity: 98+%
CAS No.: 4224-70-8
Molecular Formula: Br(CH2)5CO2H
Formula Weight: 195.06
Flash Point: 95°(203°F)
Odour: Odourless
Boiling Point: 165-170°/20mm
Beilstein Reference: 1749740
UN No.: 3261
MDL No.: MFCD00004422
Vzorec:
BrH₂C(CH₂)₄COOH Bod varu: 165…170 °C (20 mmHg) Bod tání: 32…35 °C Hustota: 1,4068 g/cm³ (30 °C) Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004422 CAS číslo: 4224-70-8 EINECS: 224-176-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...