4-Bromobutyric acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Bromobutanoic acid
A12870.09
A12870.18
A12870.30
A12870.09EA
1650
CZK
A12870.09
A12870.18
A12870.30
4-Bromobutyric acid ≥97%
4-Bromobutyric acid
Purity: 97%
CAS No.: 2623-87-2
Molecular Formula: Br(CH2)3CO2H
Formula Weight: 167.01
Flash Point: 115°(239°F)
Odour: Stench
Boiling Point: 129-131°/11mm
Beilstein Reference: 1743056
UN No.: 3261
MDL No.: MFCD00002817
Vzorec:
BrH₂C(CH₂)₂COOH MW: 167 Bod varu: 129…131 °C (11 mmHg) Bod tání: 31…33 °C Bod vzplanutí: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002817 CAS číslo: 2623-87-2 EINECS: 220-083-9 UN: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...