Methyl-5-bromovalerate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M32607-10G
M32607-25G
SIALM32607-10GEA
3670
CZK
SIALM32607-10G
SIALM32607-25G
Methyl-5-bromovalerate, Sigma-Aldrich®
Methyl-5-bromovalerate
Vzorec:
BrH₂C(CH₂)₃COOCH₃ Bod varu: 79…80 °C (5,33 hPa) Hustota: 1,36 g/cm³ (20 °C) |
MDL Number:
MFCD00000265 CAS číslo: 5454-83-1 EINECS: 226-709-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...