Tisk…

Bromoacetyl chloride, Sigma-Aldrich®

Dodavatel: Merck

16120-100ML 16120-25ML 16120-500ML
SIAL16120-100MLEA 8630 CZK
SIAL16120-100ML SIAL16120-25ML SIAL16120-500ML
Bromoacetyl chloride, Sigma-Aldrich®
Bromoacetyl chloride
Vzorec: BrCH₂COOCl
MW: 157,39 g/mol
MDL Number: MFCD00000724
CAS číslo: 22118-09-8
EINECS: 244-790-7
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR