3-Bromopropionyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Bromopropanoyl chloride
M-Clarity™ quality level = MQ100
142514-25G
142514-100G
SIAL142514-25GEA
2150
CZK
SIAL142514-25G
SIAL142514-100G
3-Bromopropionyl chloride, Sigma-Aldrich®
3-Bromopropionyl chloride
Vzorec:
BrH₂CCH₂COOCl MW: 171,42 g/mol Bod varu: 55 °C (1013 hPa) Hustota: 1,7 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000746 CAS číslo: 15486-96-1 EINECS: 239-515-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...